siRNA Details
| Gene Symbol: GTSE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016426.6 | NP_057510.4 | 10 | 1951 | ||
| XM_005261627.3 | XP_005261684.1 | 10 | 2811 | |||
| XM_011530211.2 | XP_011528513.1 | 9 | 2753 | |||
| XM_017028815.1 | XP_016884304.1 | 9 | 2201 | |||
| GenBank mRNA | AF223408.1 | - | 9 | 1752 | ||
| AK025563.1 | - | 10 | 1778 | |||
| AK222806.1 | - | 10 | 1778 | |||
| AK303087.1 | - | 9 | 1722 | |||
| AK315550.1 | - | 10 | 1777 | |||
| AY927427.1 | - | 4 | 691 | |||
| BC006325.2 | - | 9 | 1729 | 4138532 | ||
| BC075828.1 | - | 10 | 1852 | 5092448 | ||
| CF145254.1 | - | 4 | 395 | |||
| CT841519.1 | - | 9 | 1695 | |||

GTSE1
Genomic Map