siRNA Details
| Gene Symbol: CXXC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317199.1 | NP_001304128.1 | 3 | 1288 | ||
| NM_001317200.1 | NP_001304129.1 | 2 | 725 | |||
| NM_001317201.1 | NP_001304130.1 | 2 | 777 | |||
| NM_001317202.1 | NP_001304131.1 | 2 | 741 | |||
| NM_001317203.1 | NP_001304132.1 | 3 | 949 | |||
| NM_001317204.1 | NP_001304133.1 | 3 | 819 | |||
| NM_001317205.1 | NP_001304134.1 | 2 | 818 | |||
| NM_001317206.1 | NP_001304135.1 | 4 | 1045 | |||
| NM_001317207.1 | NP_001304136.1 | 2 | 727 | |||
| NM_001317208.1 | NP_001304137.1 | 3 | 833 | |||
| NM_001317209.1 | NP_001304138.1 | 2 | 739 | |||
| NM_001317210.1 | NP_001304139.1 | 3 | 827 | |||
| NM_001317211.1 | NP_001304140.1 | 3 | 821 | |||
| NM_016463.8 | NP_057547.5 | 2 | 1194 | |||
| XM_017009573.1 | XP_016865062.1 | 2 | 1064 | |||
| GenBank mRNA | AB097005.1 | - | 2 | 771 | ||
| AB097032.1 | - | 2 | 771 | |||
| AF151029.1 | - | 1 | 400 | |||
| AK001782.1 | - | 2 | 716 | |||
| AK024338.1 | - | 2 | 1194 | |||
| AK314420.1 | - | 1 | 478 | |||
| AY007103.1 | - | 1 | 839 | |||
| BC002490.2 | - | 1 | 530 | 3050465 | ||
| BC006428.1 | - | 2 | 1125 | 3343683 | ||
| BC017439.1 | - | 2 | 1086 | 2989328 | ||
| BC024040.1 | - | 2 | 1115 | 3925666 | ||
| BC041013.1 | - | 2 | 1115 | 4139961 | ||
| BC050046.1 | - | 2 | 1087 | |||
| GQ379202.1 | - | 1 | 480 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_133687, AK005364, BC016207, BC089314, AK015150, and others.

CXXC5
Genomic Map