siRNA Details
| Gene Symbol: RAB6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016577.3 | NP_057661.3 | 1, 2 | 457 | ||
| GenBank mRNA | AF166492.1 | - | 1, 2 | 413 | ||
| AF498940.1 | - | 1, 2 | 76 | |||
| AK312378.1 | - | 1, 2 | 377 | |||
| BC002510.2 | - | 1, 2 | 426 | 3050592 | ||
| BC078662.1 | - | 1, 2 | 205 | 6450986 | ||
| BT007263.1 | - | 1, 2 | 76 | |||
| DA221889.1 | - | 1, 2 | 457 | |||

RAB6B
Genomic Map