siRNA Details
| Gene Symbol: ESF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276380.1 | NP_001263309.1 | 13, 14 | 2356 | ||
| NM_016649.3 | NP_057733.2 | 13, 14 | 2367 | |||
| XM_017027874.1 | XP_016883363.1 | 13, 14 | 3819 | |||
| GenBank mRNA | AK022670.1 | - | 1, 2 | 157 | ||
| AK074299.1 | - | 12, 13 | 1853 | |||

ESF1
Genomic Map