siRNA Details
| Gene Symbol: PDHA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000284.3 | NP_000275.1 | 3 | 325 | ||
| NM_001173454.1 | NP_001166925.1 | 4 | 439 | |||
| NM_001173455.1 | NP_001166926.1 | 3 | 325 | |||
| NM_001173456.1 | NP_001166927.1 | 3 | 325 | |||
| XM_017029574.1 | XP_016885063.1 | 4 | 426 | |||
| GenBank mRNA | AK129887.1 | - | 3 | 260 | ||
| AK222740.1 | - | 3 | 256 | |||
| AK222988.1 | - | 3 | 290 | |||
| AK293250.1 | - | 4 | 404 | |||
| AK296341.1 | - | 3 | 291 | |||
| AK296457.1 | - | 3 | 239 | |||
| AK312263.1 | - | 3 | 291 | |||
| BC002406.1 | - | 3 | 237 | 2961286 | ||
| DC361191.1 | - | 3 | 325 | |||
| EF590117.1 | - | 357 | ||||
| J03503.1 | - | 3 | 252 | |||
| J03575.1 | - | 3 | 225 | |||
| L13318.1 | - | 3 | 285 | |||
| L48690.1 | - | 3 | 285 | |||
| M24848.1 | - | 3 | 223 | |||
| X52709.1 | - | 3 | 234 | |||
| X52710.1 | - | 3 | 236 | |||

PDHA1
Genomic Map