siRNA Details
| Gene Symbol: PDHA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005390.4 | NP_005381.1 | 1 | 734 | ||
| GenBank mRNA | AK313872.1 | - | 1 | 694 | ||
| BC030697.2 | - | 1 | 730 | 4824350 | ||
| BC066953.1 | - | 1 | 716 | 5273609 | ||
| BC094760.1 | - | 1 | 729 | |||
| BC119656.1 | - | 1 | 702 | 40115940 | ||
| BC119657.1 | - | 1 | 702 | 40115941 | ||
| BC127637.1 | - | 1 | 702 | 40115943 | ||
| BC127638.1 | - | 1 | 701 | 40115944 | ||

PDHA2
Genomic Map