siRNA Details
| Gene Symbol: PDK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199898.1 | NP_001186827.1 | 7, 8 | 895 | ||
| NM_001199899.1 | NP_001186828.1 | 6, 7 | 694 | |||
| NM_002611.4 | NP_002602.2 | 6, 7 | 857 | |||
| GenBank mRNA | AK055119.1 | - | 7, 8 | 895 | ||
| AK290522.1 | - | 6, 7 | 756 | |||
| AK293600.1 | - | 6, 7 | 671 | |||
| AK297089.1 | - | 6, 7 | 644 | |||
| BC005811.2 | - | 6, 7 | 745 | 3831465 | ||
| BC040478.1 | - | 6, 7 | 800 | 4690182 | ||
| BC063137.1 | - | 6, 7 | 771 | 4272593 | ||
| L42451.1 | - | 6, 7 | 744 | |||

PDK2
Genomic Map