siRNA Details
| Gene Symbol: PEX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000288.3 | NP_000279.1 | 10 | 1045 | ||
| XM_005267019.4 | XP_005267076.1 | 10 | 1325 | |||
| XM_006715502.1 | XP_006715565.1 | 7 | 1095 | |||
| XM_017010934.1 | XP_016866423.1 | 7 | 1112 | |||
| GenBank mRNA | AL583681.3 | - | 1 | 42 | ||
| BC006268.1 | - | 10 | 987 | 3948721 | ||
| BC031606.1 | - | 11 | 1137 | 5176358 | ||
| CR542054.1 | - | 10 | 943 | |||
| CR542076.1 | - | 10 | 943 | |||
| U76560.1 | - | 10 | 991 | |||
| U88871.1 | - | 10 | 1024 | |||

PEX7
Genomic Map