siRNA Details
| Gene Symbol: PI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002638.3 | NP_002629.1 | 2 | 323 | ||
| GenBank mRNA | AA583773.1 | - | 2 | 253 | ||
| BC010952.1 | - | 2 | 324 | 4083155 | ||
| CR542234.1 | - | 2 | 299 | |||
| Z18538.1 | - | 2 | 301 | |||

PI3
Genomic Map