siRNA Details
| Gene Symbol: IL17D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138284.1 | NP_612141.1 | 2, 3 | 393 | ||
| XM_005266421.3 | XP_005266478.1 | 2, 3 | 837 | |||
| GenBank mRNA | AF458062.1 | - | 2, 3 | 343 | ||
| AF479775.1 | - | 1, 2 | 285 | |||
| AY078238.1 | - | 2, 3 | 393 | |||
| AY359113.1 | - | 1, 2 | 285 | |||
| BC036243.1 | - | 2, 3 | 396 | 5301608 | ||

IL17D
Genomic Map