siRNA Details
| Gene Symbol: MBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281453.1 | NP_001268382.1 | 2 | 339 | ||
| NM_001281454.1 | NP_001268383.1 | 2 | 243 | |||
| GenBank mRNA | AF072247.1 | - | 2 | 232 | ||
| AK001474.1 | - | 1 | 39 | |||
| AK290882.1 | - | 2 | 286 | |||
| BC009372.1 | - | 2 | 175 | 4122128 | ||
| BC043619.1 | - | 2 | 193 | 6156267 | ||
| BU181763.1 | - | 2 | 339 | |||

MBD3
Genomic Map