siRNA Details
Gene Symbol: LURAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013615.2 | NP_001013633.1 | 1 | 199 | ||
GenBank mRNA | AK057892.1 | - | 1 | 199 | ||
BC034422.1 | - | 1 | 192 | 5163612 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LURAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013615.2 | NP_001013633.1 | 1 | 199 | ||
GenBank mRNA | AK057892.1 | - | 1 | 199 | ||
BC034422.1 | - | 1 | 192 | 5163612 |