siRNA Details
| Gene Symbol: GPR173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018969.5 | NP_061842.1 | 2 | 1176 | ||
| XM_011530798.2 | XP_011529100.1 | 2 | 1193 | |||
| GenBank mRNA | AB040801.1 | - | 1 | 918 | ||
| BC009861.2 | - | 2 | 1176 | 3936037 | ||
| BM670611.1 | - | 1 | 536 | |||
More Information
Set Membership: |
|

GPR173
Genomic Map