siRNA Details
Gene Symbol: GPR173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018969.5 | NP_061842.1 | 2 | 783 | ||
XM_011530798.2 | XP_011529100.1 | 2 | 800 | |||
GenBank mRNA | AB040801.1 | - | 1 | 525 | ||
BC009861.2 | - | 2 | 783 | 3936037 | ||
BI602566.1 | - | 1 | 575 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
