siRNA Details
| Gene Symbol: SLC38A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307936.1 | NP_001294865.1 | 13 | 1596 | ||
| NM_018976.4 | NP_061849.2 | 14 | 1712 | |||
| GenBank mRNA | AB037803.2 | - | 13 | 1272 | ||
| AF259799.1 | - | 13 | 1292 | |||
| AF298897.1 | - | 14 | 1623 | |||
| AJ344099.1 | - | 14 | 1624 | |||
| AK001700.1 | - | 13 | 1505 | |||
| AK172784.1 | - | 7 | 719 | |||
| AK291644.1 | - | 14 | 1416 | |||
| AK293858.1 | - | 7 | 827 | |||
| BC040342.1 | - | 14 | 1614 | 3874551 | ||
| CR457267.1 | - | 10 | 972 | |||

SLC38A2
Genomic Map