siRNA Details
| Gene Symbol: FBXO42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018994.2 | NP_061867.1 | 10 | 1760 | ||
| XM_006710698.3 | XP_006710761.1 | 9 | 1533 | |||
| XM_017001502.1 | XP_016856991.1 | 11 | 1889 | |||
| XM_017001503.1 | XP_016856992.1 | 11 | 1819 | |||
| GenBank mRNA | AB037753.1 | - | 9 | 1309 | ||
| AK055598.1 | - | 10 | 1723 | |||
| AL833874.1 | - | 1 | 454 | |||
| BC006174.1 | - | 6 | 894 | |||
| BC043410.1 | - | 10 | 1722 | 6155685 | ||
| BC063864.1 | - | 10 | 1722 | 6182628 | ||

FBXO42
Genomic Map