siRNA Details
| Gene Symbol: FAM134B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034850.2 | NP_001030022.1 | 8 | 1032 | ||
| NM_019000.4 | NP_061873.2 | 6 | 908 | |||
| XM_011514053.2 | XP_011512355.1 | 9 | 1091 | |||
| XM_011514054.2 | XP_011512356.1 | 7 | 1042 | |||
| XM_011514055.2 | XP_011512357.1 | 6 | 832 | |||
| GenBank mRNA | AK024920.1 | - | 6 | 738 | ||
| AK025808.1 | - | 8 | 959 | |||
| AL832438.1 | - | 8 | 708 | |||
| BC030517.1 | - | 2 | 104 | |||
| BC053326.1 | - | 8 | 956 | 5014868 | ||
| BC073132.1 | - | 6 | 741 | 5760145 | ||
| CR994722.1 | - | 2 | 147 | |||

FAM134B
Genomic Map