siRNA Details
| Gene Symbol: FAM64A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195228.1 | NP_001182157.1 | 2 | 377 | ||
| NM_019013.2 | NP_061886.2 | 2 | 377 | |||
| XM_011523944.2 | XP_011522246.1 | 2 | 487 | |||
| GenBank mRNA | AK001018.1 | - | 1 | 317 | ||
| AK001353.1 | - | 2 | 337 | |||
| AK225613.1 | - | 2 | 339 | |||
| AK225702.1 | - | 2 | 337 | |||
| BC005004.1 | - | 2 | 344 | 3533875 | ||
| BC013966.2 | - | 2 | 323 | 2988318 | ||
| KU178653.1 | - | 1 | 268 | |||
| KU178654.1 | - | 1 | 268 | |||

FAM64A
Genomic Map