siRNA Details
| Gene Symbol: C11orf71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019021.3 | NP_061894.2 | 2 | 583 | ||
| GenBank mRNA | BC071695.1 | - | 2 | 450 | 5416335 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C11orf71| Gene Symbol: C11orf71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019021.3 | NP_061894.2 | 2 | 583 | ||
| GenBank mRNA | BC071695.1 | - | 2 | 450 | 5416335 | |