siRNA Details
| Gene Symbol: DDIT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019058.2 | NP_061931.1 | 3 | 838 | ||
| GenBank mRNA | AF335324.1 | - | 3 | 854 | ||
| AK000507.1 | - | 3 | 833 | |||
| AL136668.1 | - | 3 | 837 | |||
| AY090097.1 | - | 3 | 754 | |||
| BC000708.1 | - | 3 | 744 | 3507244 | ||
| BC007714.1 | - | 3 | 826 | 4302878 | ||
| BC015236.1 | - | 3 | 812 | 3906374 | ||

DDIT4
Genomic Map