siRNA Details
Gene Symbol: MXRA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282582.1 | NP_001269511.1 | 4 | 415 | ||
NM_001282583.1 | NP_001269512.1 | 3 | 834 | |||
NM_001282585.1 | NP_001269514.1 | 3 | 390 | |||
NM_032348.3 | NP_115724.1 | 3 | 637 | |||
XM_017001516.1 | XP_016857005.1 | 3 | 414 | |||
GenBank mRNA | AB052096.1 | - | 4 | 415 | ||
AK027269.1 | - | 3 | 390 | |||
AK095966.1 | - | 3 | 834 | |||
AK225028.1 | - | 3 | 376 | |||
AK304615.1 | - | 3 | 374 | |||
AK315510.1 | - | 3 | 404 | |||
BC006213.2 | - | 3 | 378 | 3343261 | ||
BC017312.2 | - | 3 | 378 | 5090754 |
