siRNA Details
| Gene Symbol: FBXL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099784.2 | NP_001093254.2 | 9 | 1781 | ||
| NM_001282351.1 | NP_001269280.1 | 9 | 1649 | |||
| GenBank mRNA | AK098777.1 | - | 7 | 1310 | ||
| AK127701.1 | - | 9 | 1633 | |||
| AK302669.1 | - | 9 | 1783 | |||
| BC059173.1 | - | 8 | 1297 | 5738074 | ||

FBXL19
Genomic Map