siRNA Details
| Gene Symbol: EPDR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017549.4 | NP_060019.2 | 1, 2 | 648 | ||
| GenBank mRNA | AJ250475.2 | - | 1, 2 | 121 | ||
| AK290885.1 | - | 1, 2 | 342 | |||
| AY027862.2 | - | 1, 2 | 648 | |||
| BC000686.2 | - | 1, 2 | 392 | |||
| BC018299.1 | - | 1, 2 | 540 | 3461888 | ||
| DA938402.1 | - | 2, 3 | 161 | |||

EPDR1
Genomic Map