siRNA Details
| Gene Symbol: NUTM2F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017561.1 | NP_060031.1 | 7 | 2083 | ||
| Gene Symbol: NUTM2G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170741.1 | NP_001164212.1 | 7 | 2038 | ||

NUTM2F
Genomic Map