siRNA Details
| Gene Symbol: GRAMD1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017577.4 | NP_060047.3 | 2 | 187 | ||
| XM_005247546.2 | XP_005247603.1 | 2 | 199 | |||
| XM_005247547.1 | XP_005247604.1 | 2 | 149 | |||
| XM_011512930.1 | XP_011511232.1 | 2 | 182 | |||
| XM_017006646.1 | XP_016862135.1 | 2 | 197 | |||
| XM_017006647.1 | XP_016862136.1 | 2 | 177 | |||
| GenBank mRNA | AK097783.1 | - | 2 | 183 | ||
| AK292846.1 | - | 2 | 123 | |||
| AK293884.1 | - | 2 | 143 | |||
| BC035040.1 | - | 2 | 193 | 4829130 | ||

GRAMD1C
Genomic Map