siRNA Details
| Gene Symbol: DCHS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017639.3 | NP_060109.2 | 12, 13 | 2661 | ||
| XM_011532045.1 | XP_011530347.1 | 7, 8 | 4449 | |||
| XM_011532046.2 | XP_011530348.1 | 6, 7 | 2105 | |||
| GenBank mRNA | AY354497.1 | - | 12, 13 | 2661 | ||

DCHS2
Genomic Map