siRNA Details
| Gene Symbol: GIPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017655.5 | NP_060125.4 | 1, 2 | 435 | ||
| GenBank mRNA | AB073737.1 | - | 1, 2 | 251 | ||
| AK225941.1 | - | 1, 2 | 274 | |||
| BC036075.1 | - | 1, 2 | 435 | 5296057 | ||
| BI561167.1 | - | 1, 2 | 441 | |||

GIPC2
Genomic Map