siRNA Details
| Gene Symbol: ASPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017680.4 | NP_060150.4 | 6 | 1026 | ||
| GenBank mRNA | AF316824.1 | - | 5 | 744 | ||
| AK000136.1 | - | 4 | 396 | |||
| AK308107.1 | - | 6 | 940 | |||
| AY029191.1 | - | 6 | 959 | |||
| AY358329.1 | - | 6 | 959 | |||
| BC063114.1 | - | 6 | 991 | 30404602 | ||

ASPN
Genomic Map