siRNA Details
| Gene Symbol: ESRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034915.2 | NP_001030087.2 | 5 | 759 | ||
| NM_001122825.1 | NP_001116297.1 | 5 | 759 | |||
| NM_001122826.1 | NP_001116298.1 | 5 | 759 | |||
| NM_001122827.1 | NP_001116299.1 | 5 | 759 | |||
| NM_017697.3 | NP_060167.2 | 5 | 759 | |||
| XM_005250991.3 | XP_005251048.1 | 5 | 896 | |||
| XM_005250992.3 | XP_005251049.1 | 5 | 896 | |||
| GenBank mRNA | AK000178.1 | - | 2 | 153 | ||
| BC019932.2 | - | 5 | 720 | |||
| BC035406.1 | - | 2 | 172 | |||
| BC067098.1 | - | 5 | 713 | 6586195 | ||
| BC099916.1 | - | 5 | 723 | 30527936 | ||
| BC101621.1 | - | 2 | 118 | |||
| BC112043.1 | - | 2 | 118 | |||
| BX647570.1 | - | 5 | 821 | |||

ESRP1
Genomic Map