siRNA Details
| Gene Symbol: DNAJC25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015882.2 | NP_001015882.2 | 2 | 446 | ||
| NR_037148.1 | 3 | 684 | ||||
| GenBank mRNA | AF447177.1 | - | 3 | 653 | ||
| AK095452.1 | - | 3 | 667 | |||
| BC048318.1 | - | 2 | 417 | 4456137 | ||
| DR004317.1 | - | 2 | 453 | |||

DNAJC25
Genomic Map