siRNA Details
| Gene Symbol: DCAF16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017741.3 | NP_060211.3 | 3 | 1497 | ||
| XM_005248169.3 | XP_005248226.1 | 3 | 1485 | |||
| XM_005248171.3 | XP_005248228.1 | 2 | 1381 | |||
| XM_006713966.3 | XP_006714029.1 | 4 | 1656 | |||
| XM_017008327.1 | XP_016863816.1 | 2 | 2950 | |||
| XM_017008328.1 | XP_016863817.1 | 2 | 2935 | |||
| GenBank mRNA | AK000287.1 | - | 1 | 619 | ||
| AK056116.1 | - | 3 | 1497 | |||
| BC050697.1 | - | 2 | 1158 | 6044417 | ||
| BC068025.1 | - | 1 | 796 | 4593954 | ||
| BC101716.1 | - | 1 | 507 | 8069222 | ||
| BC101718.1 | - | 1 | 507 | 8069224 | ||

DCAF16
Genomic Map