siRNA Details
| Gene Symbol: PLPPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017753.2 | NP_060223.2 | 5, 6 | 895 | ||
| NM_207299.1 | NP_997182.1 | 5, 6 | 1028 | |||
| GenBank mRNA | AK000307.1 | - | 5, 6 | 895 | ||
| AK225306.1 | - | 5, 6 | 895 | |||
| AK303718.1 | - | 3, 4 | 775 | |||
| AK303807.1 | - | 4, 5 | 891 | |||
| AK309115.1 | - | 3, 4 | 604 | |||
| AY304515.1 | - | 5, 6 | 761 | |||
| AY337718.1 | - | 4, 5 | 641 | |||
| BC022465.1 | - | 5, 6 | 761 | 4797229 | ||

PLPPR1
Genomic Map