siRNA Details
| Gene Symbol: UHRF1BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017754.3 | NP_060224.3 | 14 | 2208 | ||
| GenBank mRNA | AB126777.1 | - | 14 | 2043 | ||
| AK122918.1 | - | 1 | 31 | |||
| BC137120.1 | - | 14 | 2038 | 9021117 | ||
| BC137127.1 | - | 14 | 2038 | 9021124 | ||
| BX640641.1 | - | 14 | 2077 | |||

UHRF1BP1
Genomic Map