siRNA Details
| Gene Symbol: WHSC1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017778.2 | NP_060248.2 | 8 | 2257 | ||
| NM_023034.1 | NP_075447.1 | 8 | 2257 | |||
| GenBank mRNA | AF255649.1 | - | 9 | 1859 | ||
| AF332468.1 | - | 8 | 2257 | |||
| AF332469.1 | - | 8 | 2257 | |||
| AJ295990.1 | - | 8 | 2052 | |||
| AJ295991.1 | - | 8 | 2052 | |||
| AJ295992.1 | - | 8 | 2052 | |||
| AK000360.1 | - | 3 | 556 | |||
| AK022560.1 | - | 8 | 1934 | |||
| BC012059.1 | - | 8 | 2225 | |||
| BC062631.1 | - | 8 | 2196 | |||
| BC073858.1 | - | 7 | 1783 | |||
| BC101717.1 | - | 8 | 1831 | 8069223 | ||
| BC113469.1 | - | 8 | 1831 | 8322521 | ||
| BC143510.1 | - | 8 | 1831 | 9052018 | ||

WHSC1L1
Genomic Map