siRNA Details
| Gene Symbol: TMEM161A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256766.1 | NP_001243695.1 | 8, 9 | 855 | ||
| NM_017814.2 | NP_060284.1 | 10, 11 | 1164 | |||
| XM_011528117.2 | XP_011526419.1 | 5, 6 | 925 | |||
| XM_017026916.1 | XP_016882405.1 | 10, 11 | 1060 | |||
| GenBank mRNA | AK000429.1 | - | 10, 11 | 1121 | ||
| AK096964.1 | - | 8, 9 | 820 | |||
| AK225424.1 | - | 10, 11 | 1078 | |||
| AY358088.1 | - | 10, 11 | 1148 | |||
| BC005210.1 | - | 10, 11 | 1124 | 4041226 | ||
| BC016478.2 | - | 9, 10 | 1088 | |||

TMEM161A
Genomic Map