siRNA Details
| Gene Symbol: FAM206A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017832.3 | NP_060302.1 | 5 | 479 | ||
| GenBank mRNA | AF153417.1 | - | 5 | 409 | ||
| AK000464.1 | - | 5 | 463 | |||
| AK310297.1 | - | 4 | 305 | |||
| BC015795.1 | - | 5 | 691 | 3910513 | ||

FAM206A
Genomic Map