siRNA Details
| Gene Symbol: NMRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127603.1 | NP_001121075.1 | 3, 4 | 375 | ||
| NM_017881.2 | NP_060351.1 | 3, 4 | 375 | |||
| NR_023352.1 | 4, 5 | 632 | ||||
| XM_006717162.1 | XP_006717225.1 | 4, 5 | 660 | |||
| XM_006717163.2 | XP_006717226.1 | 4, 5 | 695 | |||
| XM_011518817.1 | XP_011517119.1 | 3, 4 | 485 | |||
| XM_011518818.1 | XP_011517120.1 | 3, 4 | 1007 | |||
| XM_011518819.2 | XP_011517121.1 | 3, 4 | 1007 | |||
| XM_017014868.1 | XP_016870357.1 | 5, 6 | 981 | |||
| XM_017014869.1 | XP_016870358.1 | 4, 5 | 1288 | |||
| XM_017014870.1 | XP_016870359.1 | 3, 4 | 1031 | |||
| XM_017014871.1 | XP_016870360.1 | 4, 5 | 660 | |||
| XM_017014872.1 | XP_016870361.1 | 5, 6 | 981 | |||
| XM_017014873.1 | XP_016870362.1 | 5, 6 | 952 | |||
| XM_017014874.1 | XP_016870363.1 | 4, 5 | 695 | |||
| GenBank mRNA | AK000566.1 | - | 3, 4 | 340 | ||
| AK023413.1 | - | 3, 4 | 367 | |||
| AK097144.1 | - | 4, 5 | 605 | |||
| AY611480.1 | - | 2, 3 | 130 | |||
| BC001366.2 | - | 3, 4 | 283 | 3051165 | ||
| BC036804.1 | - | 3, 4 | 375 | 5727308 | ||
| CB135264.1 | - | 3, 4 | 375 | |||

NMRK1
Genomic Map