siRNA Details
| Gene Symbol: RMND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017909.3 | NP_060379.2 | 2 | 517 | ||
| XM_017010988.1 | XP_016866477.1 | 2 | 490 | |||
| GenBank mRNA | AK000634.1 | - | 2 | 460 | ||
| AK292339.1 | - | 2 | 492 | |||
| BC012081.1 | - | 1 | 354 | |||
| BC106065.1 | - | 2 | 479 | 6165385 | ||
| BC119683.1 | - | 2 | 382 | 40116343 | ||
| DA747450.1 | - | 2 | 517 | |||

RMND1
Genomic Map