siRNA Details
| Gene Symbol: PID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100818.1 | NP_001094288.1 | 2 | 347 | ||
| NM_017933.4 | NP_060403.3 | 3 | 558 | |||
| XM_011511384.2 | XP_011509686.1 | 2 | 230 | |||
| XM_017004404.1 | XP_016859893.1 | 3 | 5489 | |||
| XM_017004405.1 | XP_016859894.1 | 4 | 428 | |||
| XM_017004406.1 | XP_016859895.1 | 2 | 86 | |||
| GenBank mRNA | AB075874.1 | - | 3 | 213 | ||
| AK000708.1 | - | 2 | 186 | |||
| AK096636.1 | - | 2 | 289 | |||
| AK125359.1 | - | 3 | 482 | |||
| AY317148.1 | - | 2 | 179 | |||
| DB474424.1 | - | 2 | 347 | |||

PID1
Genomic Map