siRNA Details
| Gene Symbol: AIM1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039775.3 | NP_001034864.2 | 13 | 4205 | ||
| XM_005245918.2 | XP_005245975.1 | 13 | 4191 | |||
| XM_011541672.1 | XP_011539974.1 | 12 | 4155 | |||
| XM_011541673.2 | XP_011539975.1 | 13 | 4430 | |||
| GenBank mRNA | AK095339.1 | - | 12 | 1249 | ||
| BC136870.1 | - | 9 | 947 | 9020861 | ||

AIM1L
Genomic Map