siRNA Details
| Gene Symbol: UBR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175748.3 | NP_786924.2 | 7 | 904 | ||
| NR_038150.1 | 6 | 770 | ||||
| GenBank mRNA | AK001345.1 | - | 4 | 282 | ||
| AK097477.1 | - | 7 | 889 | |||
| AK315492.1 | - | 7 | 703 | |||
| BC015046.1 | - | 7 | 703 | 3908032 | ||
| BC017013.1 | - | 6 | 548 | 3883770 | ||
| BC051819.1 | - | 7 | 666 | 6171393 | ||
| BX248249.1 | - | 7 | 663 | |||
| DA158479.1 | - | 6 | 544 | |||

UBR7
Genomic Map