siRNA Details
| Gene Symbol: SHQ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018130.2 | NP_060600.2 | 7 | 926 | ||
| XM_011533895.2 | XP_011532197.1 | 7 | 954 | |||
| XM_011533896.2 | XP_011532198.1 | 7 | 954 | |||
| XM_011533897.2 | XP_011532199.1 | 7 | 954 | |||
| XM_011533898.2 | XP_011532200.1 | 7 | 954 | |||
| XM_011533899.1 | XP_011532201.1 | 7 | 954 | |||
| XM_017006724.1 | XP_016862213.1 | 7 | 954 | |||
| GenBank mRNA | AK001401.1 | - | 7 | 924 | ||
| AK092962.1 | - | 7 | 924 | |||
| AK296789.1 | - | 7 | 825 | |||
| BC017204.2 | - | 7 | 916 | 3617948 | ||
| BC017274.2 | - | 7 | 904 | 4840343 | ||
| BC025270.1 | - | 7 | 916 | 4622871 | ||
| BC032671.1 | - | 7 | 930 | 5538687 | ||
| BC039830.1 | - | 7 | 859 | 6064610 | ||
| BC047879.2 | - | 7 | 877 | 5541019 | ||

SHQ1
Genomic Map