siRNA Details
| Gene Symbol: DCUN1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014283.1 | NP_001014305.1 | 2 | 164 | ||
| XM_017020643.1 | XP_016876132.1 | 2 | 375 | |||
| XM_017020644.1 | XP_016876133.1 | 2 | 305 | |||
| XM_017020645.1 | XP_016876134.1 | 2 | 346 | |||
| XM_017020646.1 | XP_016876135.1 | 2 | 4087 | |||
| XM_017020647.1 | XP_016876136.1 | 2 | 625 | |||
| XM_017020648.1 | XP_016876137.1 | 2 | 1388 | |||
| GenBank mRNA | AK001566.1 | - | 2 | 147 | ||
| BC056669.1 | - | 2 | 151 | 6494926 | ||
| BM836766.1 | - | 2 | 307 | |||

DCUN1D2
Genomic Map