siRNA Details
| Gene Symbol: FANCI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113378.1 | NP_001106849.1 | 26 | 2964 | ||
| NM_018193.2 | NP_060663.2 | 25 | 2784 | |||
| XM_011521756.2 | XP_011520058.1 | 26 | 2988 | |||
| XM_011521757.2 | XP_011520059.1 | 26 | 2984 | |||
| XM_011521764.2 | XP_011520066.1 | 25 | 2808 | |||
| XM_011521767.2 | XP_011520069.1 | 26 | 2958 | |||
| GenBank mRNA | AB058697.1 | - | 12 | 2888 | ||
| AK001581.1 | - | 19 | 2173 | |||
| AK027564.1 | - | 14 | 1671 | |||
| AK055176.1 | - | 25 | 2743 | |||
| BC021859.1 | - | 3 | 306 | |||
| BC140769.1 | - | 26 | 2921 | 9021637 | ||
| BC144483.1 | - | 26 | 2921 | 9053010 | ||
| EF469766.1 | - | 25 | 2874 | |||
| EF567077.1 | - | 25 | 2874 | |||

FANCI
Genomic Map