siRNA Details
| Gene Symbol: PNMAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103149.1 | NP_001096619.1 | 2 | 327 | ||
| NM_018215.3 | NP_060685.2 | 2 | 327 | |||
| XM_011527067.2 | XP_011525369.1 | 2 | 341 | |||
| GenBank mRNA | AK290218.1 | - | 2 | 275 | ||
| AL834344.1 | - | 2 | 171 | |||
| BC032508.1 | - | 2 | 305 | 5246377 | ||
| BC051688.1 | - | 2 | 284 | 5295669 | ||
| DA219062.1 | - | 2 | 302 | |||
| DA325514.1 | - | 2 | 327 | |||

PNMAL1
Genomic Map