siRNA Details
| Gene Symbol: MRGBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018270.5 | NP_060740.1 | 1, 2 | 272 | ||
| NR_136405.1 | 1, 2 | 272 | ||||
| GenBank mRNA | AB085682.1 | - | 1, 2 | 172 | ||
| AK001776.1 | - | 1, 2 | 224 | |||
| BC009889.2 | - | 1, 2 | 191 | 4026378 | ||
| BC014235.2 | - | 1, 2 | 196 | 4555060 | ||
| BC018841.2 | - | 1, 2 | 191 | 3050866 | ||
| BM920599.1 | - | 178 | ||||
| BU902487.1 | - | 1, 2 | 272 | |||

MRGBP
Genomic Map