siRNA Details
| Gene Symbol: TMEM40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284406.1 | NP_001271335.1 | 3 | 352 | ||
| NM_001284407.1 | NP_001271336.1 | 3 | 264 | |||
| NM_018306.3 | NP_060776.2 | 3 | 264 | |||
| XM_011533937.2 | XP_011532239.1 | 4 | 519 | |||
| GenBank mRNA | AK001898.1 | - | 3 | 196 | ||
| AK092470.1 | - | 3 | 264 | |||
| AK301984.1 | - | 3 | 352 | |||
| BC020658.1 | - | 3 | 214 | 4768606 | ||

TMEM40
Genomic Map