siRNA Details
| Gene Symbol: GIMAP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018326.2 | NP_060796.1 | 3 | 854 | ||
| XM_005250017.1 | XP_005250074.1 | 3 | 870 | |||
| GenBank mRNA | AF117333.1 | - | 3 | 822 | ||
| AK001972.1 | - | 3 | 816 | |||
| AK301441.1 | - | 3 | 873 | |||
| AK301606.1 | - | 4 | 622 | |||
| BC020657.1 | - | 3 | 831 | 4701647 | ||

GIMAP4
Genomic Map