siRNA Details
| Gene Symbol: CPPED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099455.1 | NP_001092925.1 | 3 | 608 | ||
| NM_018340.2 | NP_060810.2 | 4 | 1034 | |||
| GenBank mRNA | AK002013.1 | - | 4 | 1017 | ||
| AK022710.1 | - | 4 | 984 | |||
| AK093854.1 | - | 4 | 780 | |||
| AK225359.1 | - | 4 | 984 | |||
| AK225617.1 | - | 4 | 1029 | |||
| AK298662.1 | - | 3 | 438 | |||
| AY553877.1 | - | 4 | 923 | |||
| BC006289.2 | - | 4 | 1018 | 3959701 | ||
| BX640805.1 | - | 3 | 574 | |||

CPPED1
Genomic Map