siRNA Details
| Gene Symbol: TMEM144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018342.4 | NP_060812.2 | 4 | 685 | ||
| XM_005263110.1 | XP_005263167.1 | 5 | 640 | |||
| XM_005263112.3 | XP_005263169.1 | 6 | 675 | |||
| XM_006714254.1 | XP_006714317.1 | 5 | 497 | |||
| XM_017008366.1 | XP_016863855.1 | 3 | 653 | |||
| GenBank mRNA | AK002017.1 | - | 3 | 589 | ||
| BC026318.1 | - | 4 | 380 | |||
| BC036551.1 | - | 3 | 1963 | |||
| BC048117.1 | - | 3 | 1958 | |||
| BC054487.1 | - | 4 | 433 | 5732836 | ||
| BC060875.1 | - | 4 | 411 | 4800219 | ||

TMEM144
Genomic Map